Skip to Content
Merck
All Photos(1)

Key Documents

528131

Millipore

PIPES, Free Acid, ULTROL® Grade

A zwitterionic N-substituted aminosulfonic acid useful in the pH range of 6.1-7.5. Has a pKa 6.76 at 25°C. Absorbance (0.5%, 0.25 N NaOH, 260 nm):0.18.

Synonym(s):

Piperazine-N,Nʹ- bis(2-ethanesulfonic Acid), Piperazine-N,Nʹ-bis(2-ethanesulfonic Acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O6S2
CAS Number:
Molecular Weight:
302.37
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% dry basis (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

pKa (20 °C)

6.8

solubility

aqueous buffer: 50 mg/mL
water: soluble

cation traces

heavy metals: <10 ppm

shipped in

ambient

storage temp.

15-25°C

InChI

1S/C8H18N2O6S2.Na/c11-17(12,13)7-5-9-1-2-10(4-3-9)6-8-18(14,15)16;/h1-8H2,(H,11,12,13)(H,14,15,16);/q;+1/p-1

InChI key

OGGAIRCLBMGXCZ-UHFFFAOYSA-M

General description

A zwitterionic N-substituted aminosulfonic acid useful in the pH range of 6.1-7.5. Has a pKa 6.76 at 25°C.
Absorbance (0.5%, 0.25 N NaOH, 260 nm): <0.18.
Zwitterionic N-substituted aminosulfonic acid, active in the pH range of 6.0-8.5. Has a pKa of 6.8 at 20°C.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 3 months at 4°C.





From Catalog:

Desc. Field- replaced "active in the pH range 6.0-8.5"

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service