1.93686
Potassium hexacyanoferrate(II) trihydrate
EMPLURA®
Synonym(s):
Potassium hexacyanoferrate(II) trihydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
product line
EMPLURA®
storage temp.
room temp
InChI
1S/6CN.Fe.4K.3H2O/c6*1-2;;;;;;;;/h;;;;;;;;;;;3*1H2/q6*-1;+2;4*+1;;;
InChI key
UTYXJYFJPBYDKY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service