Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249C2C7

J & W Pharmlab LLC

2-Trimethylsilanyl-propionitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
127.26
MDL number:

Iupac Name

2-(trimethylsilyl)propanenitrile

Inchi Code

1S/C6H13NSi/c1-6(5-7)8(2,3)4/h6H,1-4H3

InChI key

WISPPBDNOQXVLN-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Physical Form

Colorless Liquid

Product Link

https://www.jwpharmlab.com/home/product/05R0517

MSDS

https://www.jwpharmlab.com/home/product/05R0517


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service