Skip to Content
Merck
All Photos(1)

Key Documents

ATEH98C4728E

AstaTech, Inc.

4-METHOXYBENZYL CARBONOCHLORIDATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
200.62
MDL number:

Pricing and availability is not currently available. Contact Technical Service

Iupac Name

4-methoxybenzyl carbonochloridate

Inchi Code

1S/C9H9ClO3/c1-12-8-4-2-7(3-5-8)6-13-9(10)11/h2-5H,6H2,1H3

InChI key

BOVISBAPSYSQPI-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Sticky Oil to Semi-Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=201739

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=D81342


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service