Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AMBH95E08066

Ambeed, Inc.

2-(Oxetan-3-yl)acetic acid

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
116.12
MDL number:

Iupac Name

3-oxetanylacetic acid

Inchi Code

1S/C5H8O3/c6-5(7)1-4-2-8-3-4/h4H,1-3H2,(H,6,7)

InChI key

FSJPCGQVHNWRGI-UHFFFAOYSA-N

storage temp.

Sealed in dry,Store in freezer, under -20C

Purity

98%

Country of Origin

CN

Shipping Temperature

Cold-chain transportation

Physical Form

solid

Product Link

https://www.ambeed.com/products/1310381-54-4.html

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.ambeed.com/products/1310381-54-4.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library