Skip to Content
Merck
All Photos(1)

Key Documents

W371203

Sigma-Aldrich

Methyl trans-2-octenoate

≥96%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CHCO2CH3
CAS Number:
Molecular Weight:
156.22
FEMA Number:
3712
EC Number:
Council of Europe no.:
11800
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.299
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥96%

refractive index

n20/D 1.442 (lit.)

bp

89-91 °C/9 mmHg (lit.)

density

0.898 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; fruity

SMILES string

[H]\C(CCCCC)=C(\[H])C(=O)OC

InChI

1S/C9H16O2/c1-3-4-5-6-7-8-9(10)11-2/h7-8H,3-6H2,1-2H3/b8-7+

InChI key

CJBQSBZJDJHMLF-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mengya Wang et al.
Applied microbiology and biotechnology, 102(19), 8511-8524 (2018-08-09)
Persisters represent a small bacterial population that is dormant and that survives under antibiotic treatment without experiencing genetic adaptation. Persisters are also considered one of the major reasons for recalcitrant chronic bacterial infections. Although several mechanisms of persister formation have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service