Skip to Content
Merck
All Photos(1)

Documents

W343501

Sigma-Aldrich

3-Methyl-2-cyclopentenone

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C5H5(=O)
CAS Number:
Molecular Weight:
96.13
Beilstein:
1280476
MDL number:
Flavis number:
7.112

biological source

synthetic

Assay

≥97%

refractive index

n20/D 1.488 (lit.)

bp

74 °C/15 mmHg (lit.)

mp

3-5 °C (lit.)

density

0.980 g/mL at 20 °C
0.971 g/mL at 25 °C (lit.)

SMILES string

CC1=CC(=O)CC1

InChI

1S/C6H8O/c1-5-2-3-6(7)4-5/h4H,2-3H2,1H3

InChI key

CHCCBPDEADMNCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Sweet, fruity, fatty, stinging, chemical

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service