Skip to Content
Merck
All Photos(1)

Documents

V1708

Sigma-Aldrich

9-Vinylanthracene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12
CAS Number:
Molecular Weight:
204.27
Beilstein:
2041641
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

97%

bp

61-66 °C/10 mmHg (lit.)

mp

62-65 °C (lit.)

storage temp.

2-8°C

SMILES string

C=Cc1c2ccccc2cc3ccccc13

InChI

1S/C16H12/c1-2-14-15-9-5-3-7-12(15)11-13-8-4-6-10-16(13)14/h2-11H,1H2

InChI key

OGOYZCQQQFAGRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceren Kütahya et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(46), 10444-10451 (2020-04-29)
NIR exposure at 790 nm activated photopolymerization of monomers comprising UV-absorbing moieties by using [CuII /(TPMA)]Br2 (TPMA=tris(2-pyridylmethyl)amine) in the ppm range and an alkyl bromide as initiator. Some of them comprised structural elements selected either from those showing proton transfer or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service