Skip to Content
Merck
All Photos(1)

Documents

447919

Sigma-Aldrich

2,5-Bis(5-tert-butyl-benzoxazol-2-yl)thiophene

98%

Synonym(s):

BBOT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26N2O2S
CAS Number:
Molecular Weight:
430.56
Beilstein:
631803
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

mp

199-201 °C (lit.)

SMILES string

CC(C)(C)c1ccc2oc(nc2c1)-c3ccc(s3)-c4nc5cc(ccc5o4)C(C)(C)C

InChI

1S/C26H26N2O2S/c1-25(2,3)15-7-9-19-17(13-15)27-23(29-19)21-11-12-22(31-21)24-28-18-14-16(26(4,5)6)8-10-20(18)30-24/h7-14H,1-6H3

InChI key

AIXZBGVLNVRQSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in poly bottles

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service