Skip to Content
Merck
All Photos(1)

Key Documents

394653

Supelco

N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane

with 10% trimethylchlorosilane, for GC derivatization, ≥98.0%

Synonym(s):

BSTFA + TMCS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C[=NSi(CH3)3]OSi(CH3)3
CAS Number:
Molecular Weight:
257.40
Beilstein:
2050024
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

for GC derivatization

Assay

≥98.0%
98% (excluding TMCS)

quality

with 10% trimethylchlorosilane

contains

10% TMCS

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

refractive index

n20/D 1.384 (lit.)

bp

45-50 °C/14 mmHg (lit.)

density

0.961 g/mL at 25 °C
0.97 g/mL (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)O\C(=N\[Si](C)(C)C)C(F)(F)F

InChI

1S/C8H18F3NOSi2/c1-14(2,3)12-7(8(9,10)11)13-15(4,5)6/h1-6H3/b12-7+

InChI key

XCOBLONWWXQEBS-KPKJPENVSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

The position of the silyl groups has not been unequivocally established. It has been referred to as both N,N and N,O.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

15.8 °F - closed cup

Flash Point(C)

-9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hidenori Matsukami et al.
The Science of the total environment, 493, 672-681 (2014-07-06)
Chlorinated and non-halogenated organophosphorus flame retardants (OPFRs) including tris(2-chloroisopropyl) phosphate (TCIPP), diethylene glycol bis(di(2-chloroisopropyl) phosphate) (DEG-BDCIPP), triphenyl phosphate (TPHP), and bisphenol A bis(diphenyl phosphate) (BPA-BDPP) have been used increasingly as alternatives to polybrominated diphenyl ethers and other brominated flame retardants.
Ahmed I Rushdi et al.
SpringerPlus, 3, 253-253 (2014-06-14)
Propolis is a sticky material mixed by honeybees to utilize it in protecting their hives from infection by bacteria and fungi. The therapeutic properties of propolis are due to its chemical composition with bio-active compounds; therefore, researchers are interested in
Nyi Nyi Naing et al.
Journal of chromatography. A, 1423, 1-8 (2015-11-05)
Electro membrane extraction-solid-liquid phase microextraction (EME-SLPME) was developed for the first time to determine phenolic contaminants in water. The extraction system consisted of a solid/liquid interface that permitted a three-phase microextraction approach involving an aqueous sample (donor phase): an organic
Greg M Kowalski et al.
Biochemical and biophysical research communications, 462(1), 27-32 (2015-05-02)
Defects in muscle glucose metabolism are linked to type 2 diabetes. Mechanistic studies examining these defects rely on the use of high fat-fed rodent models and typically involve the determination of muscle glucose uptake under insulin-stimulated conditions. While insightful, they
Fabrício Flores-Nunes et al.
Environmental science and pollution research international, 22(22), 17267-17279 (2014-11-16)
The composition and concentration of substances in urban effluents are complex and difficult to measure. These contaminants elicit biological responses in the exposed organisms. Proteomic analysis is a powerful tool in environmental toxicology by evidencing alterations in protein expression due

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service