Increase Your Throughput
Samplicity® G2 Filtration System
Samplicity® G2 Filtration System
For scientists who filter dozens of samples a day for HPLC and LC-MS analyses, traditional filtration can lead to manual fatigue — especially for hard-to-filter, viscous, or particulate-laden samples. Our vacuum-driven Samplicity® G2 filtration system enables scientists to skip the pain of manual filtration. With multiple sample ports, researchers can filter up to eight samples simultaneously into standard 2 mL HPLC sample vials, saving you time in the lab.
More Samples. Less Time.
- Faster than manual filtration: use the Samplicity® G2 system to filter up to eight samples at once into standard 2 mL HPLC vials
- Simplify filtration with adapter funnels for loading samples into Millex® 33mm filters. Millex® for Samplicity® G2 eliminates the need for syringes when using Millex® syringe filters.
- Handle difficult-to-filter samples: increase efficiency when working with samples containing high particulate levels
Reduced Waste
The Samplicity® G2 system reduces the amount of liquid and solid waste generated and eliminates segregation of syringe waste.
| Samplicity® G2 system | Syringe filtration | % Reduction in Waste |
---|---|---|---|
Weight (g) | 2827.5 | 4232.9 | 33.2% |
Volume (cubic inches) | 1000 | 3000 | 66% |
Fifteen users filtered 24 samples. Syringes, syringe filters, and Millex Samplicity® membrane filters were collected, weighed, and placed into 10 in x 10 in x 10 in boxes to evaluate relative waste volume.
Samplicity® G2 Filtration System (Bold Blue)
Samplicity® G2 Filtration System (Glossy Green)
Low extractable Millex Samplicity® Filters for LC-MS
Trusted Millex® filter quality
For chromatographers who filter dozens of samples a day, Millex® for Samplicity® G2 simplifies workflows with nonsterile 33mm Millex® filters by adding an adapter funnel to load samples. Designed with the needs of sample preparation in mind, Millex® filters for Samplicity® G2 feature:
- Broad offering of membrane types for sample and solvent compatibility
- 0.20, 0.22, and 0.45 µm pore sizes
- Solvent-resistant, polypropylene resin used for Millex® filter housing and funnels
Millex® Filters for sample preparation