Merck
HomePhotometry & ReflectometrySucrose in Soft Drinks
Sucrose in Soft Drinks
Reflectometric determination after enzymatic conversion.
Sugar (sucrose) cubes
Introduction
Chemical analysis of foods and beverages is performed to ensure safe consumption, product quality, and product integrity. As a part of quality control, soft drink producers must monitor the concentration of sugar (sucrose) in their final products and during the manufacturing process. In this application note, we describe a method to determine sucrose in soft drinks with the Reflectoquant® Sucrose (Saccharose) Test and RQflex® 20 reflectometer.
Experimental Method
Sucrose (saccharose) is cleaved by sucrose phosphorylase into fructose and glucose-1-phosphate. The latter is converted by phosphoglucomutase into glucose-6-phosphate. This is in turn oxidized by NAD under the catalytic effect of glucose-6-phosphate dehydrogenase to gluconate-6-phosphate. In the presence of diaphorase the NADH formed in the process reduces a tetrazolium salt to a blue formazan that is determined reflectometrically.
Reagents, Instruments and Materials
Test Kits
  • Sucrose (Saccharose) Test Method: reflectometric with test strips and reagent 0.25 - 2.50 g/l Reflectoquant® (1.16141)
Instruments
  • Reflectometer RQflex® 20 Reflectoquant® (1.17246)*
*This application note pertains to RQflex® 20 and all discontinued instruments (RQflex® 10, RQflex® plus).
Sample preparation
Dilute the sample depending on the anticipated sucrose content, e. g. for a 1:100 dilution, pipette 0.5 ml of sample into a 50-ml volumetric flask and make up to the mark with distilled water. The measurement results should preferably lie in the middle of the measuring range. It may be necessary to repeat the dilution step with a further dilution.
Instrumental Analysis
  • Place 10 ml of distilled water, 5 drops of reagent Sa-1, and 1 ml of correspondingly diluted sample in the test glass and swirl.
  • Press the start button of the reflectometer and at the same time immerse the test strip with both reaction zones into the measurement sample (20 - 26 °C) for approx. 2 seconds.
  • Allow excess liquid to run off over the long edge of the strip onto an absorbent paper towel.
  • Immediately insert the test strip into the strip adapter.
  • The test strip is measured in the reflectometer after 300 seconds.
The measurement value [g/l] is saved automatically. (See the operating manual for RQflex® and the instruction for use for Reflectoquant® Sucrose (Saccharose) Test.)
Serial measurement
After the first measurement, any number of further measurements can be made by pressing the start button. This, however, requires the use of a stopwatch since the countdown function of the reflectometer is available only for the first measurement. The detection zones of the test strip must be appropriately protected from light during the reaction time.
Calculation
Sucrose content [g/l] = measurement value [g/l] x dilution factor
Results
Table 1.Measurement values for cola (Nominal conc. in cola acc. to manufacturer: 10.6 g/100 ml. Result HPLC: 10.5 g/100 ml)
Table 2.Measurement values for orangeade (Nominal conc. in orangeade acc. to manufacturer 9.4 g/100 ml)