Merck
LC/MS Analysis of Conjugated Bile Acids on Ascentis® Express RP-Amide
LC/MS Analysis of Conjugated Bile Acids on Ascentis® Express RP-Amide application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express RP-Amide, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 5 cm × 4.6 mm
mobile phase component
Product No.
Description
Pricing
Ammonium acetate
LiChropur, eluent additive for LC-MS
standard
Product No.
Description
Pricing
Taurocholic acid sodium salt hydrate
≥95% (HPLC)
Sodium glycochenodeoxycholate
Sodium taurochenodeoxycholate
Sodium taurodeoxycholate hydrate
≥95% (HPLC)
Glycocholic acid hydrate
synthetic, ≥97% (HPLC)
CONDITIONS
column
Ascentis Express RP-Amide, 5 cm x 4.6 mm I.D., 2.7 μm particles (53922-U)
mobile phase
[A] 10 mM ammonium acetate, pH 3 with acetic acid; [B] acetonitrile
gradient
25 to 40% B in 10 min
flow rate
1.0 mL/min (postcolumn split to the MS)
column temp.
35 °C
detector
MS, ESI(-), SIR
injection
5 μL
sample
10 mg/L in mobile phase A
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany