HPLC Analysis of 5-Methyl-5-Phenylhydantoin Enantiomers on Astec® CHIROBIOTIC® TAG (Fast Flow)
![HPLC Analysis of 5-Methyl-5-Phenylhydantoin Enantiomers on Astec® CHIROBIOTIC® TAG (Fast Flow) application for HPLC](/deepweb/assets/sigmaaldrich/product/images/117/653/70753ecb-961f-48a0-89b3-31c87a8a60e5/800/70753ecb-961f-48a0-89b3-31c87a8a60e5.jpg)
CONDITIONS
column
CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 μm particles (14024AST)
mobile phase
methanol
flow rate
0.8 mL/min
column temp.
23 °C
detector
UV, 220 nm
injection
2 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC