HPLC Analysis of 15N-Acetylcysteine Enantiomers on Astec® (R,R) P-CAP™
![HPLC Analysis of 15N-Acetylcysteine Enantiomers on Astec® (R,R) P-CAP™ application for HPLC](/deepweb/assets/sigmaaldrich/product/images/317/079/49b150c1-efcb-4fc9-8936-2c86593303c1/800/49b150c1-efcb-4fc9-8936-2c86593303c1.jpg)
CONDITIONS
column
Astec (R,R) P-CAP, 25 cm x 4.6 mm I.D., 5 μm particles (31024AST)
mobile phase
20 mM ammonium acetate in: (A) acetonitrile; (B) methanol; (70:30, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
1 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
P-CAP is a trademark of Sigma-Aldrich Co. LLC