GC Analysis of Ionic Liquid Test Mix on SLB®-IL76 (30 m x 0.25 mm I.D., 0.20 μm)
![GC Analysis of Ionic Liquid Test Mix on SLB®-IL76 (30 m x 0.25 mm I.D., 0.20 μm) suitable for GC](/deepweb/assets/sigmaaldrich/product/images/212/802/c1f12ac2-e50f-4ebc-ac06-ab67b5f97914/800/c1f12ac2-e50f-4ebc-ac06-ab67b5f97914.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-IL76, 30 m x 0.25 mm I.D., 0.20 μm (28913-U)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 26 cm/sec
injection
1.0 μL 100:1 split
sample
Ionic Liquid Test Mix #1 (N09000), each analyte at various concentrations in isooctane
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany