GC Analysis of Glycols and Diols on Equity-1
![GC Analysis of Glycols and Diols on Equity-1 suitable for GC](/deepweb/assets/sigmaaldrich/product/images/177/301/352fd106-7882-4957-b232-dc84066e007f/800/352fd106-7882-4957-b232-dc84066e007f.jpg)
CONDITIONS
column
Equity-1, 30 m x 0.53 mm I.D., 0.50 μm (28068-U)
oven
50 °C (2 min), 8 °C/min to 200 °C (2 min)
inj. temp.
250 °C
detector
FID, 300 °C
carrier gas
helium, 37 cm/sec, constant
injection
1 μL, pulsed (10 psi until 0.10 min) splitless (0.75 min)
liner
2 mm I.D., straight
sample
each analyte at 100 ppm in methanol
Description
Legal Information
null