GC Analysis of Background Levels of a Tenax® TA Thermal Desorption Tube on SPB®-1
![GC Analysis of Background Levels of a Tenax® TA Thermal Desorption Tube on SPB®-1 suitable for GC](/deepweb/assets/sigmaaldrich/product/images/147/974/cf47038f-8fd6-41f9-bffb-791a93417589/800/cf47038f-8fd6-41f9-bffb-791a93417589.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SPB-1, 60 m x 0.25 mm I.D., 1.0 μm (24031)
oven
35 °C (8 min), 5 °C/min to 100 °C, 15 °C/min to 230 °C (hold 4 min)
inj. temp.
200 °C
MSD interface
230 °C
carrier gas
nitrogen, 2 mL/min. constant
injection
split 10:1
Description
Legal Information
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany
Tenax is a registered trademark of Buchem B.V.