GC Analysis of Alcohol Enantiomers on β-DEX™ 325
CONDITIONS
column
β-DEX 325, 30 m × 0.25 mm I.D., 0.25 μm (24308)
oven
110 °C, 1-phenyl-2-propanol; 100 °C, isopinocampheol; 90 °C, 6-methyl-5-hepten-2-ol
inj. temp.
220 °C
detector
FID, 300 °C
carrier gas
helium, 20 cm/sec @ analysis temperature
injection
1 μL 100:1 split
sample
1 mg/mL in methylene chloride
Description
Legal Information
DEX is a trademark of Sigma-Aldrich Co. LLC