GC Analysis of a 16-Component Anilines Mix on SLB®-IL59
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-IL59, 30 m x 0.25 mm I.D., 0.20 mm (28891-U)
oven
75 °C, (2 min) 10 °C/min to 200 °C, 20 °C/min to 300 °C (10 min)
inj. temp.
250 °C
detector
FIID, 300 °C
carrier gas
helium, 1.5 mL/min constant
injection
0.5 μL 100:1 split
liner
4 mm I.D., split/splitless type, single taper design
sample
Each analyte at 5000 μg/mL in methylene chloride
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany