Merck
  • Comprehensive two-dimensional gas chromatography of the 209 polychlorinated biphenyls.
Comprehensive two-dimensional gas chromatography of the 209 polychlorinated biphenyls.
Journal of chromatography. A (2003-12-03)
Mikael Harju, Conny Danielsson, Peter Haglund
ABSTRACT
Comprehensive two-dimensional gas chromatography (GC x GC) of the 209 polychlorinated biphenyls (CBs) was carried out using a longitudinally modulated cryogenic system (LMCS) and liquid carbon dioxide as cryogen. The effluent from a non-polar column was modulated and further separated on either a polar or a shape-selective second-dimension column. Five GC x GC column combinations were evaluated, with DB-XLB as the first column in each case. DB-XLB separates more congeners than any other GC column currently available. When combined with a biscyanopropyl siloxane (SP-2340 or BPX70) or smectic liquid crystal (LC-50) second-dimension column in a GC x GC system many additional CBs can be separated. In total, 176 and 181 of the 209 congeners were separated (Rs = 0.5) on the column combinations DB-XLB/SP-2340 and DB-XLB/LC-50, respectively. Of the 136 CBs present in any Aroclor mixture at concentrations greater than 0.05% (w/w), 126 were resolved using either of these two column combinations. The seven frequently measured CBs 28, 52, 101, 118, 138, 153, 180, and the WHO-PCBs 77, 81, 105, 114, 118, 123, 126, 156. 157, 167, 169 and 189 were all separated from Aroclor CBs on the DB-XLB/LC-50 column set, whereas CBs 118 and 131 coeluted on the DB-XLB/SP-2340 column set. In addition, three technical CB formulations (Aroclors 1232, 1248 and 1260) and a seal blubber sample (Halichoerus grypus) from the Baltic Sea were analyzed. Similar peak patterns were found for Aroclor 1260 and the CBs in the seal blubber, facilitating use of this technical CB formulation to identify the CBs in the blubber by GC x GC. Individual CBs in environmental samples, such as seal blubber, may be identified semi-automatically by matching the samples GC x GC profiles to a template generated using a standard containing all 209 CBs. Using such a template, 64 CBs were identified in the grey seal blubber sample.
Product Description
Supelco
SP®-2340 Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 0.20 μm