Merck
  • A general and efficient copper catalyst for the amidation of aryl halides.
A general and efficient copper catalyst for the amidation of aryl halides.
Journal of the American Chemical Society (2002-06-20)
Artis Klapars, Xiaohua Huang, Stephen L Buchwald
ABSTRACT
An experimentally simple and inexpensive catalyst system was developed for the amidation of aryl halides by using 0.2-10 mol % of CuI, 5-20 mol % of a 1,2-diamine ligand, and K(3)PO(4), K(2)CO(3), or Cs(2)CO(3) as base. Catalyst systems based on N,N'-dimethylethylenediamine or trans-N,N'-dimethyl-1,2-cyclohexanediamine were found to be the most active even though several other 1,2-diamine ligands could be used in the easiest cases. Aryl iodides, bromides, and in some cases even aryl chlorides can be efficiently amidated. A variety of functional groups are tolerated in the reaction, including many that are not compatible with Pd-catalyzed amidation or amination methodology.
Sigma-Aldrich
4,7-Dimethoxy-1,10-phenanthroline, 97%