- Identification of proteins with the CDw75 epitope in human colorectal cancer.
Identification of proteins with the CDw75 epitope in human colorectal cancer.
The CDw75 epitope is an α(2,6) sialylated antigen overexpressed in colorectal cancer (CRC), where its expression correlates with the progression of the disease. The CDw75 epitope is located mainly in N-glycoproteins, whose identity remains unknown. The aim of the present study was to identify proteins with the CDw75 epitope as a strategy to deepen the understanding of molecular pathogenesis of CRC and to identify novel biomarkers for this disease. For this purpose, a two-dimensional electrophoresis approach was employed. Protein spots in the gels were matched to the corresponding CDw75 positive spots in the immunoblotted polyvinylidene difluoride membranes, and further identification of the protein species was performed by mass spectrometry. Additionally, one-dimensional western blotting experiments were performed to verify the expression of these candidate proteins in the colorectal tissue and their coincidence in molecular mass with the CDw75-positive bands. The findings of the present study indicate that haptoglobin and the keratins 8 (K8) and 18 (K18) are proteins with the CDw75 epitope in the colorectal tissue from CRC patients and also suggest novel functions and cellular locations for these proteins in the colorectal tissue and in relation to CRC.