Merck
  • IL-33 is induced in undifferentiated, non-dividing esophageal epithelial cells in eosinophilic esophagitis.
IL-33 is induced in undifferentiated, non-dividing esophageal epithelial cells in eosinophilic esophagitis.
Scientific reports (2017-12-16)
J Travers, M Rochman, J M Caldwell, J A Besse, C E Miracle, M E Rothenberg
ABSTRACT
The molecular and cellular etiology of eosinophilic esophagitis (EoE), an emerging tissue-specific allergic disease, involves dysregulated gene expression in esophageal epithelial cells. Herein, we assessed the esophageal expression of IL-33, an epithelium-derived alarmin cytokine, in patients with EoE. IL-33 protein was markedly overexpressed within the nuclei of a subpopulation of basal layer esophageal epithelial cells in patients with active EoE compared to control individuals. IL-33 exhibited dynamic expression as levels normalized upon EoE remission. IL-33-positive basal epithelial cells expressed E-cadherin and the undifferentiated epithelial cell markers keratin 5 and 14 but not the differentiation marker keratin 4. Moreover, the IL-33-positive epithelial cells expressed the epithelial progenitor markers p75 and p63 and lacked the proliferation markers Ki67 and phospho-histone H3. Additionally, the IL-33-positive cells had low expression of PCNA. IL-33 expression was detected in ex vivo-cultured primary esophageal epithelial cells in a subpopulation of cells lacking expression of proliferation markers. Collectively, we report that IL-33 expression is induced in an undifferentiated, non-dividing esophageal epithelial cell population in patients with active EoE.