Merck
  • The Epithelial Circumferential Actin Belt Regulates YAP/TAZ through Nucleocytoplasmic Shuttling of Merlin.
The Epithelial Circumferential Actin Belt Regulates YAP/TAZ through Nucleocytoplasmic Shuttling of Merlin.
Cell reports (2017-08-10)
Kana T Furukawa, Kazunari Yamashita, Natsuki Sakurai, Shigeo Ohno
ABSTRACT
Circumferential actin belts underlying the adherens junctions of columnar epithelial cell monolayers control intercellular surface tension and cell shape to maintain tissue integrity. Yes-associated protein (YAP) and its paralog TAZ are proliferation-activating transcriptional coactivators that shuttle between the nucleus and cytoplasm. Previous studies suggest the importance of stress fibers in the actin cytoskeleton for regulation of YAP nuclear localization; however, the role of the circumferential actin belt on YAP localization remains unclarified. By manipulating actin tension, we demonstrate that circumferential actin belt tension suppresses YAP/TAZ nuclear localization. This suppression requires Merlin, an F-actin binding protein associated with adherens junctions. Merlin physically interacts with YAP/TAZ, and nuclear export sequences of Merlin are required for suppression. Together, with the observation that the association between E-cadherin and Merlin was diminished by tension in circumferential actin belts, our results suggest that released Merlin undergoes nucleocytoplasmic shutting and mediates export of YAP/TAZ from the nucleus.
Sigma-Aldrich
Anti-Myosin IIB, non-muscle antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Leptomycin B solution from Streptomyces sp., ≥95% (HPLC), Supplied in methanol: water (7:3)
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
(−)-Blebbistatin, solid, synthetic