- Mutations in IFT-A satellite core component genes
Mutations in IFT-A satellite core component genes
Cilia (2017-04-13)
Ivan Duran, S Paige Taylor, Wenjuan Zhang, Jorge Martin, Faisal Qureshi, Suzanne M Jacques, Robert Wallerstein, Ralph S Lachman, Deborah A Nickerson, Michael Bamshad, Daniel H Cohn, Deborah Krakow
PMID28400947
ABSTRACT
Skeletal ciliopathies comprise a spectrum of ciliary malfunction disorders that have a profound effect on the skeleton. Most common among these disorders is short rib polydactyly syndrome (SRPS), a recessively inherited perinatal lethal condition characterized by a long narrow chest, markedly shortened long bones, polydactyly and, often, multi-organ system involvement. SRPS shows extensive locus heterogeneity with mutations in genes encoding proteins that participate in cilia formation and/or function. Herein we describe mutations in This study defines a new
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid