Merck
  • Generation of Monoclonal Antibodies against Immunoglobulin Proteins of the Domestic Ferret (
Generation of Monoclonal Antibodies against Immunoglobulin Proteins of the Domestic Ferret (
Journal of immunology research (2017-03-14)
Greg A Kirchenbaum, Ted M Ross
ABSTRACT
The domestic ferret (
Sigma-Aldrich
Anti-Mouse IgG (γ-chain specific) antibody produced in goat, affinity isolated antibody, lyophilized powder