Merck
PRKCI promotes immune suppression in ovarian cancer.
Genes & development (2017-07-13)
Sharmistha Sarkar, Christopher A Bristow, Prasenjit Dey, Kunal Rai, Ruth Perets, Alejandra Ramirez-Cardenas, Shruti Malasi, Emmet Huang-Hobbs, Monika Haemmerle, Sherry Y Wu, Michael McGuire, Alexei Protopopov, Shan Jiang, Joyce F Liu, Michelle S Hirsch, Qing Chang, Alexander J Lazar, Anil K Sood, Ronny Drapkin, Ronald DePinho, Giulio Draetta, Lynda Chin
ABSTRACT
A key feature of high-grade serous ovarian carcinoma (HGSOC) is frequent amplification of the 3q26 locus harboring
Sigma-Aldrich
Anti-Tumor Necrosis Factor-α antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture