Merck
  • p53 performs an essential role in mediating the oncogenic stimulus triggered by loss of expression of neurofibromatosis type 2 during
p53 performs an essential role in mediating the oncogenic stimulus triggered by loss of expression of neurofibromatosis type 2 during
Oncology letters (2017-08-10)
Xiye Li, Hongsai Chen, Lu Xue, Xiuhong Pang, Xiaoman Zhang, Zhengjie Zhu, Weidong Zhu, Zhaoyan Wang, Hao Wu
ABSTRACT
The loss of the tumor suppressor
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF594 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution