Merck
  • Photobiomodulation with 670 nm light ameliorates Müller cell-mediated activation of microglia and macrophages in retinal degeneration.
Photobiomodulation with 670 nm light ameliorates Müller cell-mediated activation of microglia and macrophages in retinal degeneration.
Experimental eye research (2017-09-11)
Yen-Zhen Lu, Riccardo Natoli, Michele Madigan, Nilisha Fernando, Kartik Saxena, Riemke Aggio-Bruce, Haihan Jiao, Jan Provis, Krisztina Valter
ABSTRACT
Müller cells, the supporting cells of the retina, play a key role in responding to retinal stress by releasing chemokines, including CCL2, to recruit microglia and macrophages (MG/MΦ) into the damaged retina. Photobiomodulation (PBM) with 670 nm light has been shown to reduce inflammation in models of retinal degeneration. In this study, we aimed to investigate whether 670 nm light had an effect on Müller cell-initiated inflammation under retinal photo-oxidative damage (PD) in vivo and in vitro. Sprague-Dawley rats were pre-treated with 670 nm light (9J/cm