- Runx3 programs CD8
Runx3 programs CD8
Nature (2017-12-07)
J Justin Milner, Clara Toma, Bingfei Yu, Kai Zhang, Kyla Omilusik, Anthony T Phan, Dapeng Wang, Adam J Getzler, Toan Nguyen, Shane Crotty, Wei Wang, Matthew E Pipkin, Ananda W Goldrath
PMID29211713
ABSTRACT
Tissue-resident memory CD8