Merck
  • Lipoteichoic acid deficiency permits normal growth but impairs virulence of Streptococcus pneumoniae.
Lipoteichoic acid deficiency permits normal growth but impairs virulence of Streptococcus pneumoniae.
Nature communications (2017-12-14)
Nathalie Heß, Franziska Waldow, Thomas P Kohler, Manfred Rohde, Bernd Kreikemeyer, Alejandro Gómez-Mejia, Torsten Hain, Dominik Schwudke, Waldemar Vollmer, Sven Hammerschmidt, Nicolas Gisch
ABSTRACT
Teichoic acid (TA), a crucial cell wall constituent of the pathobiont Streptococcus pneumoniae, is bound to peptidoglycan (wall teichoic acid, WTA) or to membrane glycolipids (lipoteichoic acid, LTA). Both TA polymers share a common precursor synthesis pathway, but differ in the final transfer of the TA chain to either peptidoglycan or a glycolipid. Here, we show that LTA exhibits a different linkage conformation compared to WTA, and identify TacL (previously known as RafX) as a putative lipoteichoic acid ligase required for LTA assembly. Pneumococcal mutants deficient in TacL lack LTA and show attenuated virulence in mouse models of acute pneumonia and systemic infections, although they grow normally in culture. Hence, LTA is important for S. pneumoniae to establish systemic infections, and TacL represents a potential target for antimicrobial drug development.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Cell dissociation sieve - tissue grinder kit, tissue grinder homogenizer kit, stainless steel, autoclavable, 1/kit