Merck
  • Factor Xa inhibition by rivaroxaban in the trough steady state can significantly reduce thrombin generation.
Factor Xa inhibition by rivaroxaban in the trough steady state can significantly reduce thrombin generation.
British journal of clinical pharmacology (2017-09-10)
Shigeo Horinaka, Rie Sugawara, Yutaka Yonezawa, Toshihiko Ishimitsu
ABSTRACT
The aim of the present study was to demonstrate evidence of reduced thrombin generation at the trough plasma rivaroxaban concentration. A single-centre, prospective, nonrandomized, drug-intervention, self-controlled study was conducted in 51 anticoagulation therapy-naïve patients with nonvalvular atrial fibrillation. Plasma rivaroxaban concentration was measured by liquid chromatography tandem mass spectrometry (LC-MS/MS) and the anti-factor Xa chromogenic assay. Partial thrombin time (PT), protein C activity, and protein S antigen, prothrombin fragment 1 + 2 (F1 + 2), D-dimer, thrombomodulin (TM), thrombin-antithrombin complex (TAT), plasminogen activator inhibitor-1 (PAI-1) and tissue factor pathway inhibitor (TFPI) levels were also measured at the trough steady state after 4 weeks of rivaroxaban treatment and compared with baseline. Plasma concentrations obtained by the LC-MS/MS and anti-Xa assays were correlated (r = 0.841, P < 0.001). The mean concentration of rivaroxaban at the trough steady state was 23.6 ng ml Residual plasma rivaroxaban at the trough steady state may explain the antithrombin effect of rivaroxaban in patients with nonvalvular atrial fibrillation.
Sigma-Aldrich
2-Bromo-1-ethyl-pyridinium tetrafluoroborate, ≥97.0% (T)