Merck
  • Determination of benzotriazoles and benzothiazoles in human urine by UHPLC-TQMS.
Determination of benzotriazoles and benzothiazoles in human urine by UHPLC-TQMS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2017-11-02)
Jiufeng Li, Hongzhi Zhao, Yanqiu Zhou, Shunqing Xu, Zongwei Cai
ABSTRACT
Benzotriazole (BTR) and benzothiazole (BTH) derivatives are extensively applied in industrial processes and consumer products, and are thus frequently detected in the environmental matrices. Due to their potential estrogenic effects reported in animal studies, the assessment of human exposure to BTRs and BTHs is important. In this study, a method was developed and validated to determine six BTRs and five BTHs in human urine by using ultra-high performance liquid chromatography coupled with a triple quadrupole mass spectrometry (UHPLC-TQMS) in positive electrospray ionization mode. After de-conjugation by β-glucuronidase, urine samples were liquid-liquid extracted for the measurement of total concentrations of BTRs and BTHs. The linearity, detection limit, precision, recovery, accuracy and matrix effect were evaluated. The limits of detection were less than 0.5ng/mL. The validated method demonstrated good precision (RSD%<15%), linearity (R
Product Description
Supelco
4-Methyl-1H-benzotriazole, analytical standard
Sigma-Aldrich
Benzotriazole, ReagentPlus®, 99%
Sigma-Aldrich
2-Hydroxybenzothiazole, 98%