Merck
  • Triazole phenotypes and genotypic characterization of clinical Aspergillus fumigatus isolates in China.
Triazole phenotypes and genotypic characterization of clinical Aspergillus fumigatus isolates in China.
Emerging microbes & infections (2017-12-07)
Shuwen Deng, Lili Zhang, Yanfeng Ji, Paul E Verweij, Kin Ming Tsui, Ferry Hagen, Jos Houbraken, Jacque F Meis, Parida Abliz, Xiaodong Wang, Jingjun Zhao, Wanqing Liao
ABSTRACT
This study investigated the triazole phenotype and genotypic of clinical Aspergillus fumigatus isolates from China. We determined the triazole susceptibility profiles of 159 A. fumigatus isolates collected between 2011 and 2015 from four different areas in China tested against 10 antifungal drugs using the Clinical Laboratory Standard Institute M38-A2 method. For the seven itraconazole-resistant A. fumigatus isolates identified in the study, the cyp51A gene, including its promoter region, was sequenced and the mutation patterns were characterized. The resistant isolates were genotyped by microsatellite typing to determine the genetic relatedness to isolates from China and other countries. The frequency of itraconazole resistance in A. fumigatus isolates in our study was 4.4% (7/159). Six of the seven triazole-resistant isolates were recovered from the east and southeast of China, and one from was recovered from the west of China. No resistant isolates were found in the north. Three triazole-resistant isolates exhibited the TR
Product Description
Supelco
Posaconazole, VETRANAL®, analytical standard