Merck
  • Role of HSPA1L as a cellular prion protein stabilizer in tumor progression via HIF-1α/GP78 axis.
Role of HSPA1L as a cellular prion protein stabilizer in tumor progression via HIF-1α/GP78 axis.
Oncogene (2017-08-02)
J H Lee, Y-S Han, Y M Yoon, C W Yun, S P Yun, S M Kim, H Y Kwon, D Jeong, M J Baek, H J Lee, S-J Lee, H J Han, S H Lee
ABSTRACT
The cellular prion protein (PrP
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)