- Activity-Dependent Facilitation of Ca
Activity-Dependent Facilitation of Ca
The Journal of neuroscience : the official journal of the Society for Neuroscience (2017-10-19)
Giriraj Sahu, Hadhimulya Asmara, Fang-Xiong Zhang, Gerald W Zamponi, Ray W Turner
PMID29038242
ABSTRACT
Ca