Merck
  • 8-Aminoguanosine Exerts Diuretic, Natriuretic, and Glucosuric Activity via Conversion to 8-Aminoguanine, Yet Has Direct Antikaliuretic Effects.
8-Aminoguanosine Exerts Diuretic, Natriuretic, and Glucosuric Activity via Conversion to 8-Aminoguanine, Yet Has Direct Antikaliuretic Effects.
The Journal of pharmacology and experimental therapeutics (2017-09-21)
Edwin K Jackson, Zaichuan Mi
ABSTRACT
8-Aminoguanosine induces diuresis, natriuresis, glucosuria, and antikaliuresis. These effects could be mediated via 8-aminoguanosine's metabolism to 8-aminoguanine. In this study, we tested this hypothesis in anesthetized rats. First, we demonstrated that at 55- to 85-minutes post-i.v. administration, 8-aminoguanosine and 8-aminoguanine (33.5
Sigma-Aldrich
8-Aminoguanine, ≥95% (HPLC)
Sigma-Aldrich
8-Aminoguanosine, ≥98% (HPLC)
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)