Merck
  • Effects of PKM2 Gene Silencing on the Proliferation and Apoptosis of Colorectal Cancer LS-147T and SW620 Cells.
Effects of PKM2 Gene Silencing on the Proliferation and Apoptosis of Colorectal Cancer LS-147T and SW620 Cells.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2017-07-27)
Ran Ao, Lin Guan, Ying Wang, Jia-Ni Wang
ABSTRACT
This paper aims to explore the effects of pyruvate kinase (PK) M2 gene silencing on the proliferation and apoptosis of colorectal cancer (CRC) LS-147T and SW620 cells. CRC LS-147T and SW620 cells highly expressing PKM2 were randomly selected by quantitative real-time polymerase chain reaction (qRT-PCR) and then assigned into the blank (no transfection), PKM2-shRNA (transfection with shRNA) and empty plasmid (transfection with empty plasmid) groups. Immunofluorescence was applied to detect PKM2 protein expression. qRT-PCR and Western blotting were conducted to assess mRNA and protein expression of PKM2, p53 and p21. The cell counting kit-8 (CCK-8) assay was used to assess cell proliferation. Flow cytometry was used to assess the cell cycle and apoptosis rate, and a senescence-associated β-galactosidase staining kit was used to assess cell senescence. PKM2 exhibited high mRNA expression among CRC LS-147T and SW620 cells with remarkable protein expression noted in the cytoplasm and nucleus. The PKM2-shRNA group exhibited reduced PKM2 mRNA and protein expression, whereas p53 and p21 expression was increased compared with the blank and empty plasmid groups. Cell proliferation in PKM2-shRNA cells decreased significantly compared with the blank group and empty plasmid groups. The PKM2-shRNA group exhibited more cells in the G1 phase and fewer cells in the G2/M phase compared with the blank and empty plasmid groups. In addition, the PKM2-shRNA group exhibited significantly increased apoptosis rates and β-galactosidase activity compared with the blank and empty plasmid groups. Our study demonstrates that PKM2 gene silencing suppresses proliferation and promotes apoptosis in LS-147T and SW620 cells.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder