Merck
Overexpression of
Frontiers in plant science (2017-11-09)
Yinhua Ma, Yan Zhao, Xinxin Shangguan, Shaojie Shi, Ya Zeng, Yan Wu, Rongzhi Chen, Aiqing You, Lili Zhu, Bo Du, Guangcun He
ABSTRACT
It has been reported that the receptor-like cytoplasmic kinases (RLCKs) regulate many biological processes in plants, but only a few members have been functionally characterized. Here, we isolated a rice gene encoding