Merck
  • APR-246 (PRIMA-1(MET)) strongly synergizes with AZD2281 (olaparib) induced PARP inhibition to induce apoptosis in non-small cell lung cancer cell lines.
APR-246 (PRIMA-1(MET)) strongly synergizes with AZD2281 (olaparib) induced PARP inhibition to induce apoptosis in non-small cell lung cancer cell lines.
Cancer letters (2016-03-16)
Christophe Deben, Filip Lardon, An Wouters, Ken Op de Beeck, Jolien Van den Bossche, Julie Jacobs, Nele Van Der Steen, Marc Peeters, Christian Rolfo, Vanessa Deschoolmeester, Patrick Pauwels
ABSTRACT
APR-246 (PRIMA-1(Met)) is able to bind mutant p53 and restore its normal conformation and function. The compound has also been shown to increase intracellular ROS levels. Importantly, the poly-[ADP-ribose] polymerase-1 (PARP-1) enzyme plays an important role in the repair of ROS-induced DNA damage. We hypothesize that by blocking this repair with the PARP-inhibitor AZD2281 (olaparib), DNA damage would accumulate in the cell leading to massive apoptosis. We observed that APR-246 synergistically enhanced the cytotoxic response of olaparib in TP53 mutant non-small cell lung cancer cell lines, resulting in a strong apoptotic response. In the presence of wild type p53 a G2/M cell cycle block was predominantly observed. NOXA expression levels were significantly increased in a TP53 mutant background, and remained unchanged in the wild type cell line. The combined treatment of APR-246 and olaparib induced cell death that was associated with increased ROS production, accumulation of DNA damage and translocation of p53 to the mitochondria. Out data suggest a promising targeted combination strategy in which the response to olaparib is synergistically enhanced by the addition of APR-246, especially in a TP53 mutant background.
Sigma-Aldrich
PRIMA-1Met, ≥98% (HPLC)