Merck
  • Formation and degradation of nitrogen mustard-induced MGMT-DNA crosslinking in 16HBE cells.
Formation and degradation of nitrogen mustard-induced MGMT-DNA crosslinking in 16HBE cells.
Toxicology (2017-07-20)
Jin Cheng, Feng Ye, Guorong Dan, Yuanpeng Zhao, Jiqing Zhao, Zhongmin Zou
ABSTRACT
N-methyl-2,2-di(chloroethyl)amine (HN2) is a kind of bifunctional alkyltating agent, which can react with nucleophilic groups in DNA and/or protein to form HN2-bridged crosslinking of target molecules, such as DNA-protein crosslinkings (DPC). O