Merck
A CTLA-4 Antagonizing DNA Aptamer with Antitumor Effect.
Molecular therapy. Nucleic acids (2017-09-18)
Bo-Tsang Huang, Wei-Yun Lai, Yi-Chung Chang, Jen-Wei Wang, Shauh-Der Yeh, Emily Pei-Ying Lin, Pan-Chyr Yang
ABSTRACT
The successful translation of cytotoxic T lymphocyte antigen-4 (CTLA-4) blockade has revolutionized the concept of cancer immunotherapy. Although monoclonal antibody therapeutics remain the mainstream in clinical practice, aptamers are synthetic oligonucleotides that encompass antibody-mimicking functions. Here, we report a novel high-affinity CTLA-4-antagonizing DNA aptamer (dissociation constant, 11.84 nM), aptCTLA-4, which was identified by cell-based SELEX and high-throughput sequencing. aptCTLA-4 is relatively stable in serum, promotes lymphocyte proliferation, and inhibits tumor growth in cell and animal models. Our study demonstrates the developmental pipeline of a functional CTLA-4-targeting aptamer and suggests a translational potential for aptCTLA-4.