Merck
  • Andrographolide inhibits arrhythmias and is cardioprotective in rabbits.
Andrographolide inhibits arrhythmias and is cardioprotective in rabbits.
Oncotarget (2017-10-06)
Mengliu Zeng, Wanzhen Jiang, Youjia Tian, Jie Hao, Zhenzhen Cao, Zhipei Liu, Chen Fu, Peihua Zhang, Jihua Ma
ABSTRACT
Andrographolide has a protective effect on the cardiovascular system. To study its cardic-electrophysiological effects, action potentials and voltage-gated Na
Sigma-Aldrich
E-4031, ≥98% (HPLC), lyophilized powder
Sigma-Aldrich
Chromanol 293B, ≥98% (HPLC), powder