Merck
Eplerenone Prevents Atrial Fibrosis via the TGF-β Signaling Pathway.
Cardiology (2017-06-02)
Lili Du, Mu Qin, Yi Yi, Xiaoqing Chen, Weifeng Jiang, Li Zhou, Daoliang Zhang, Kai Xu, Yiqing Yang, Chao Li, Yan Liu, Xu Liu, Sheng-Zhong Duan
ABSTRACT
Eplerenone (EPL), an antagonist of the mineralocorticoid receptor, is beneficial for atrial fibrillation and atrial fibrosis. However, the underlying mechanism remains less well known. We aimed to investigate the effect of EPL on atrial fibrosis using a mouse with selective atrial fibrosis and to explore the underlying mechanisms. EPL-treated MHC-TGFcys33ser transgenic mice that have selective atrial fibrosis (Tx+EPL mice), as well as control mice, were used for in vivo studies including histological analyses, Western blotting, and qRT-PCR studies. TGF-β1-stimulated atrial fibroblasts were treated with EPL or vehicle for the in vitro studies including Western blotting and qRT-PCR studies. In addition, Smad7 siRNA was used to knock down Smad7. EPL inhibited atrial fibrosis in the Tx mice. In addition, EPL suppressed the expression of fibrosis-related molecules induced by TGF-β1 in vivo and in vitro. This occurred in concert with a downregulation of Smad7 protein expression and an upregulation of p-Smad2/3 protein expression. In addition, knockdown of Smad7 by siRNA abolished the protective roles of EPL. EPL inhibited atrial fibrosis in Tx mice. The underlying mechanism may involve increased protein expression of Smad7, which enhances the inhibitory feedback regulation of TGF-β1/Smad signaling.