Merck
  • Doxorubicin-induced mitophagy contributes to drug resistance in cancer stem cells from HCT8 human colorectal cancer cells.
Doxorubicin-induced mitophagy contributes to drug resistance in cancer stem cells from HCT8 human colorectal cancer cells.
Cancer letters (2016-12-04)
Chen Yan, Lan Luo, Chang-Ying Guo, Shinji Goto, Yoshishige Urata, Jiang-Hua Shao, Tao-Sheng Li
ABSTRACT
Cancer stem cells (CSCs) are known to be drug resistant. Mitophagy selectively degrades unnecessary or damaged mitochondria by autophagy during cellular stress. To investigate the potential role of mitophagy in drug resistance in CSCs, we purified CD133
Sigma-Aldrich
MISSION® esiRNA, targeting human ATG7