Merck
  • Oxidation of a Dimethoxyhydroquinone by Ferrihydrite and Goethite Nanoparticles: Iron Reduction versus Surface Catalysis.
Oxidation of a Dimethoxyhydroquinone by Ferrihydrite and Goethite Nanoparticles: Iron Reduction versus Surface Catalysis.
Environmental science & technology (2017-07-12)
Lelde Krumina, Gry Lyngsie, Anders Tunlid, Per Persson
ABSTRACT
Hydroquinones are important mediators of electron transfer reactions in soils with a capability to reduce Fe(III) minerals and molecular oxygen, and thereby generating Fenton chemistry reagents. This study focused on 2,6-dimethoxy hydroquinone (2,6-DMHQ), an analogue to a common fungal metabolite, and its reaction with ferrihydrite and goethite under variable pH and oxygen concentrations. Combined wet-chemical and spectroscopic analyses showed that both minerals effectively oxidized 2,6-DMHQ in the presence of oxygen. Under anaerobic conditions the first-order oxidation rate constants decreased by one to several orders of magnitude depending on pH and mineral. Comparison between aerobic and anaerobic results showed that ferrihydrite promoted 2,6-DMHQ oxidation both via reductive dissolution and heterogeneous catalysis while goethite mainly caused catalytic oxidation. These results were in agreement with changes in the reduction potential (E
Sigma-Aldrich
2,6-Dimethoxy-1,4-benzoquinone, 97%