Merck
  • Sources of S-adenosyl-L-homocysteine background in measuring protein arginine N-methyltransferase activity using tandem mass spectrometry.
Sources of S-adenosyl-L-homocysteine background in measuring protein arginine N-methyltransferase activity using tandem mass spectrometry.
Analytical biochemistry (2009-09-08)
Ted M Lakowski, Adam Frankel
ABSTRACT
S-Adenosyl-L-homocysteine (AdoHcy) background signal in reactions with protein arginine N-methyltransferase 1 is investigated using an ultrahigh-performance liquid chromatography tandem mass spectrometry assay that measures AdoHcy. We identify three sources of AdoHcy background: enzymatic automethylation, AdoHcy contamination in commercial S-adenosyl-L-methionine (AdoMet), and nonenzymatic pseudo-first-order formation of AdoHcy from AdoMet. We propose a potential mechanism for the nonenzymatic production of AdoHcy and illustrate strategies for mitigating background AdoHcy that can be applied to any assay.
Sigma-Aldrich
S-(5′-Adenosyl)-L-methionine chloride dihydrochloride, ≥75%