Merck
  • Potential L-Type Voltage-Operated Calcium Channel Blocking Effect of Drotaverine on Functional Models.
Potential L-Type Voltage-Operated Calcium Channel Blocking Effect of Drotaverine on Functional Models.
The Journal of pharmacology and experimental therapeutics (2016-10-16)
Zoltán Patai, András Guttman, Endre G Mikus
ABSTRACT
Drotaverine is considered an inhibitor of cyclic-3',5'-nucleotide-phophodiesterase (PDE) enzymes; however, published receptor binding data also support the potential L-type voltage- operated calcium channel (L-VOCC) blocking effect of drotaverine. Hence, in this work, we focus on the potential L-VOCC blocking effect of drotaverine by using L-VOCC-associated functional in vitro models. Accordingly, drotaverine and reference agents were tested on KCl-induced guinea pig tracheal contraction. Drotaverine, like the L-VOCC blockers nifedipine or diltiazem, inhibited the KCl-induced inward Ca
Sigma-Aldrich
Drotaverine hydrochloride, ≥95% (HPLC)