Merck
  • Activation of Peroxisome Proliferator-Activated Receptor Gamma and Disruption of Progesterone Synthesis of 2-Ethylhexyl Diphenyl Phosphate in Human Placental Choriocarcinoma Cells: Comparison with Triphenyl Phosphate.
Activation of Peroxisome Proliferator-Activated Receptor Gamma and Disruption of Progesterone Synthesis of 2-Ethylhexyl Diphenyl Phosphate in Human Placental Choriocarcinoma Cells: Comparison with Triphenyl Phosphate.
Environmental science & technology (2017-03-11)
Wenxin Hu, Fumei Gao, Hong Zhang, Youhei Hiromori, Shuhei Arakawa, Hisamitsu Nagase, Tsuyoshi Nakanishi, Jianying Hu
ABSTRACT
2-Ethylhexyl diphenyl phosphate (EHDPP), an organophosphate flame retardant (OPFR), is frequently detected in human blood. In this study, the sensitive dual-luciferase reporter gene assay and molecular docking were used to investigate the activation of EHDPP to human peroxisome proliferator-activated receptor gamma (PPARG). Results show that EHDPP exhibited stronger PPARG activation (EC
Sigma-Aldrich
MISSION® esiRNA, targeting human PPARG
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)